missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIPC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113428
This item is not returnable.
View return policy
Description
CIPC Polyclonal Antibody for Western Blot, IHC (P), ELISA
CIPC (CLOCK Interacting Pacemaker) is associated with Ectrodactyly, Ectodermal Dysplasia, and Cleft Lip/Palate Syndrome 1. Among its related pathways are Circadian rythm related genes.
Specifications
| CIPC | |
| Polyclonal | |
| Unconjugated | |
| CIPC | |
| 2310044G17Rik; AI842544; AI853744; Cipc; CLOCK interacting pacemaker; CLOCK interacting protein, circadian; CLOCK-interacting circadian protein; CLOCK-interacting pacemaker; CLOCK-interacting protein, circadian; KIAA1737; mKIAA1737 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 217732, 85457 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8R0W1, Q9C0C6 | |
| CIPC | |
| Recombinant Human CLOCK-interacting pacemaker protein (1-399AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction