missing translation for 'onlineSavingsMsg'
Learn More

Cipargamin, MedChemExpress

Product Code. 16436466
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436466

Brand: Medchem Express HY14430

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cipargamin (NITD609) is an potent antimalarial compound, with an IC50 of appr 1 nM against P. falciparum.

Cipargamin (NITD609) is an potent antimalarial compound, with an IC50 of appr 1 nM against P. falciparum.

Specifications

Chemical Name or Material Cipargamin
Physical Form Solid
CAS 1193314-23-6
Color Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H14Cl2FN3O
For Use With (Application) COVID-19-immunoregulation
Synonym NITD609 KAE609
Solubility Information DMSO : 50 mg/mL (128.13 mM; Need ultrasonic)
SMILES FC1=C(Cl)C=C(NC2=C3C[C@H](C)N[C@@]2(C4=C(C=CC(Cl)=C4)N5)C5=O)C3=C1
Molecular Weight (g/mol) 390.24
Formula Weight 390.24
Percent Purity 98.3%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.