missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIPAR-1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97050AF700
This item is not returnable.
View return policy
Description
CIPAR-1 Polyclonal specifically detects CIPAR-1 in Human samples. It is validated for ELISA.
Specifications
| CIPAR-1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25849 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 700 | |
| Castration-induced prostatic apoptosis-related protein 1, CIPAR-1, Prostatic androgen-repressed message 1 protein, Protein PARM-1 [Precursor] | |
| Produced in rabbits immunized with purified, recombinant CIPAR-1 (AAH13294.1; Met1-Ser258). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction