missing translation for 'onlineSavingsMsg'
Learn More

Cip4 Rabbit anti-Human, Clone: S01-6B7, Novus Biologicals™

Product Code. 18333174 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
100 ul
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333174 25 μL 25µL
18392247 100 ul 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18333174 Supplier Novus Biologicals Supplier No. NBP31501325UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cip4 Monoclonal antibody specifically detects Cip4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cip4
Applications Western Blot
Classification Monoclonal
Clone S01-6B7
Conjugate Unconjugated
Dilution Western Blot
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias cdc42-interacting protein 4, CIP4STOT, HSTP, Protein Felic, Salt tolerant protein, salt tolerator, STPTRIP-10, thyroid hormone receptor interactor 10, thyroid receptor interacting protein 10, Thyroid receptor-interacting protein 10, TR-interacting protein 10
Host Species Rabbit
Immunogen A synthetic peptide of human Cip4 (Uniprot#: Q15642)
Purification Method Affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9322
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.