missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cip4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Cip4 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227415
|
Novus Biologicals
NBP3-38370-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226680
|
Novus Biologicals
NBP3-38370-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cip4 Polyclonal antibody specifically detects Cip4 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| Cip4 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 9322 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| cdc42-interacting protein 4, CIP4STOT, HSTP, Protein Felic, Salt tolerant protein, salt tolerator, STPTRIP-10, thyroid hormone receptor interactor 10, thyroid receptor interacting protein 10, Thyroid receptor-interacting protein 10, TR-interacting protein 10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 246-545 of human Cip4 (NP_004231.1).,, Sequence:, AKCLEGMKVAANAVDPKNDSHVLIELHKSGFARPGDVEFEDFSQPMNRAPSDSSLGTPSDGRPELRGPGRSRTKRWPFGKKNKTVVTEDFSHLPPEQQRKRLQQQLEERSRELQKEVDQREALKKMKDVYEKTPQMGDPASLEPQIAETLSNIERLKLEVQKYEAWLAEAESRVLSNRGDSLSRHARPPDPPASAPPDSSSNSASQDTKESSEEPPSEESQDTPIYTEFDEDFEEEPTSPIGHCVAIYHFEGSSEGTISMAEGEDLSLMEEDKGDGWTRVRRKEGGEGYVPTSYLRVTLN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title