missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIP2A Antibody (2G10), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-59722APCCY7
This item is not returnable.
View return policy
Description
CIP2A Monoclonal specifically detects CIP2A in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry, Knockdown Validated.
Specifications
| CIP2A | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CIP2A | |
| A synthetic peptide made to a C-terminal portion of the human CIP2A protein. [UniProt No. Q8TCG1] | |
| 0.1 mL | |
| Primary | |
| p90 autoantigen (2G10) | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry, KnockDown | |
| 2G10 | |
| Flow Cytometry, KnockDown Validated | |
| Cancerous inhibitor of PP2A, CIP2A, FLJ12850, KIAA1524, MGC163436, p90, p90 autoantigen, protein CIP2A | |
| Mouse | |
| Protein G purified | |
| Cancer, Tumor Suppressors | |
| 57650.0 | |
| Human, Mouse, Rat, Primate | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction