missing translation for 'onlineSavingsMsg'
Learn More

Steris™ CIP 100™

Product Code. 15282808
Change view
Click to view available options
Volume (Metric):
3.8 L
Unit Size:
15.2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15282808 3.8 L 15.2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15282808 Supplier Steris™ Supplier No. 1D1008
  • ONLINE EXCLUSIVE PRICE
    £750.07
    £396.67 / 15.2 L
    Save £353.40
    47% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated to meet the unique cleaning demands in industries requiring an effective, validatable detergent. Steris Corporation CIP 100™ is a proprietary blend of potassium hydroxide, an advanced surfactant and other performance-enhancing ingredients that provide multiple cleaning mechanisms.

CIP 100 Alkaline Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in clean-in-place (CIP), clean-out-of-place (COP) and manual applications.

  • Flexibility—Effective cleaning against a wide variety of pharmaceutical process residues
  • Easy to Validate—Facilitates cleaning validation with a comprehensive support documentation package
  • Cost Savings—Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Global Availability—Manufactured with biodegradable surfactants to support harmonization of cleaning processes
  • Reliability—Manufactured for the pharmaceutical industry in a cGMP-compliant, FDA-registered and ISO 9001-certified facility

Additional specifications:

  • 1.27g/ml at 25°C
  • Completely soluble in water
  • Flash Point: >96.7°C
  • No phosphates
TRUSTED_SUSTAINABILITY

Specifications

pH 12.3 to 12.8 (1%)
Appearance Clear to Light Yellow
Certifications/Compliance ISO 9001, cGMP
Composition 10–30% potassium hydroxide, 1–5% tetrasodium EDTA
Format Liquid mixture
Quantity 4 Pk.
Volume (Metric) 3.8 L
compliance-icons compliance-icons
Product Identifier
  • CIP 100 - Alkaline Process and Research Cleaner
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.