missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CINPA 1

Product Code. 18732739
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732739 50 mg 50mg
18701378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18732739 Supplier Tocris Bioscience™ Supplier No. 5605/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent CAR antagonist

CINPA 1 is a potent CAR antagonist (IC50 = 70 nM). Exhibits >90-fold selectivity for CAR over pregnane X receptor (PXR). Also selective over a panel of other nuclear receptors including FXR, LXR, PPARγ and RXR.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl [5-[(diethylamino)acetyl]-10,11-dihydro-5H-dibenz[b,f]azepin-3-yl]carbamate
CAS 102636-74-8
Quantity 50 mg
Target Constitutive Androstane Receptor Antagonists
Molecular Formula C23H29N3O3
Formula Weight 395.49
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.