missing translation for 'onlineSavingsMsg'
Learn More

Cinoctramide, MedChemExpress

Product Code. 16408668
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16408668 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408668 Supplier Medchem Express Supplier No. HYB1084
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cinoctramide is an intermediate of pharmaceutical synthesis. Cinoctramide can be used for the research of autoimmune diseases.

Cinoctramide is an intermediate of pharmaceutical synthesis. Cinoctramide can be used for the research of autoimmune diseases.

Specifications

Chemical Name or Material Cinoctramide
CAS 28598-08-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H27NO4
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (374.90 mM; Need ultrasonic)
SMILES O=C(N1CCCCCCC1)/C=C/C2=CC(OC)=C(OC)C(OC)=C2
Molecular Weight (g/mol) 333.42
Formula Weight 333.42
Percent Purity 99.87%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.