missing translation for 'onlineSavingsMsg'
Learn More

Cinobufotalin, TRC

Product Code. 30010227
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010227 10 mg 10mg
30011340 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010227 Supplier Toronto Research Chemicals Supplier No. TRCC47550310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1108-68-5
Molecular Formula C26 H34 O7
Molecular Weight (g/mol) 458.54
Synonym (3β,5β,15β,16β)-16-(acetyloxy)-14,15-epoxy-3,5-dihydroxy-bufa-20,22-dienolide
IUPAC Name [(1R,2S,4R,5R,6R,7R,10S,11R,14S,16S)-14,16-dihydroxy-7,11-dimethyl-6-(6-oxopyran-3-yl)-3-oxapentacyclo[8.8.0.02,4.02,7.011,16]octadecan-5-yl] acetate
SMILES CC(=O)O[C@H]1[C@H]2O[C@]23[C@@H]4CC[C@]5(O)C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]3(C)[C@H]1C6=COC(=O)C=C6

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H34O7/c1-14(27)32-21-20(15-4-5-19(29)31-13-15)24(3)10-7-17-18(26(24)22(21)33-26)8-11-25(30)12-16(28)6-9-23(17,25)2/h4-5,13,16-18,20-22,28,30H,6-12H2,1-3H3/t16-,17-,18+,20-,21+,22+,23+,24+,25-,26+/m0/s1
Formula Weight 458.2304
Chemical Name or Material Cinobufotalin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.