missing translation for 'onlineSavingsMsg'
Learn More

Cinobufotalin, MedChemExpress

Product Code. 16461568
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461568 5 mg 5mg
16459158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461568 Supplier Medchem Express Supplier No. HYN0880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cinobufotalin is a cardiotonic steroids or bufadienolides, is extracted from the skin secretions of the giant toads. Cinobufotalin has been used as a cardiotonic, diuretic and a hemostatic agent, Cinobufotalin is also a potential anti-lung cancer agent.

Cinobufotalin is a cardiotonic steroids or bufadienolides, is extracted from the skin secretions of the giant toads. Cinobufotalin has been used as a cardiotonic, diuretic and a hemostatic agent, Cinobufotalin is also a potential anti-lung cancer agent.

Specifications

Chemical Name or Material Cinobufotalin
CAS 1108-68-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H34O7
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (272.60 mM; Need ultrasonic)
SMILES C[C@]([C@@H](C(C=C1)=COC1=O)[C@H]2OC(C)=O)(CC[C@@]3([H])[C@@]4([H])CC[C@@]5(O)[C@@]3(CC[C@H](O)C5)C)[C@@]64[C@@H]2O6
Molecular Weight (g/mol) 458.54
Formula Weight 458.54
Percent Purity 98.07%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.