missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine, TRC

Product Code. 17934085
Change view
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17934085 100 mg 100mg
17902565 5 g 5g
17971965 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17934085 Supplier Toronto Research Chemicals Supplier No. TRCC465300100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 298-57-7
Molecular Formula C26 H28 N2
Molecular Weight (g/mol) 368.51
Synonym Cinnarizine, (E)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine, Piperazine, 1-(diphenylmethyl)-4-(3-phenyl-2-propenyl)- (9CI), Piperazine, 1-cinnamyl-4-(diphenylmethyl)- (6CI,7CI,8CI), 1-(3-Phenylallyl)-4-(diphenylmethyl)piperazine, 1-Benzhydryl-4-cinnamylpiperazine, 1-Cinnamyl-4-(diphenylmethyl)piperazine, 1-Cinnamyl-4-benzhydrylpiperazine, 1-Diphenylmethyl-4-cinnamoylpiperazine, 516MD, Aplactan, Aplexal, Apotomin, Artate, Carecin, Cerebolan, Cerepar, Cinaperazine, Cinarizine, Cinazyn, Cinnacet, Cinnageron, Cinnarizine, Cinnipirine, Corathiem, Denapol, Dimitron, Dimitronal, Eglen, Folcodal, Giganten, Glanil, Hilactan, Ixterol, Katoseran, Labyrin, Lazeta, Marisan, Midronal, Mitronal, N-Benzhydryl-N'-cinnamylpiperazine, Olamin, Processine, R 1575, R 516, Sedatromin, Sepan, Siptazin, Spaderizine, Stugeron, Stunarone, Stutgeron, Stutgin, Toliman, Vergo
IUPAC Name 1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
SMILES C(\C=C\c1ccccc1)N2CCN(CC2)C(c3ccccc3)c4ccccc4

Specifica

Quantity 100 mg
InChI Formula InChI=1S/C26H28N2/c1-4-11-23(12-5-1)13-10-18-27-19-21-28(22-20-27)26(24-14-6-2-7-15-24)25-16-8-3-9-17-25/h1-17,26H,18-22H2/b13-10+
Formula Weight 368.2252
Percent Purity >95%
Chemical Name or Material Cinnarizine
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.