missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl chloride, 95%, stab. with sodium carbonate

Product Code. 11467027
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467027 250 g 250g
11457027 50 g 50g
11447027 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467027 Supplier Thermo Scientific Alfa Aesar Supplier No. A14408.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2687-12-9 | C9H9Cl | 152.62 g/mol

Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-12-9

Specifications

Melting Point 7°C
Density 1.096
Boiling Point 86°C to 87°C (2 mmHg)
Flash Point 79°C (174°F)
Molecular Formula C16H34S
Refractive Index 1.5845
MDL Number MFCD00009566
Quantity 250 g
UN Number UN2927
Beilstein 741974
Synonym dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane
Solubility Information Soluble in water (0.2g/L at 20°C).
InChI Key LOXRGHGHQYWXJK-UHFFFAOYSA-N
SMILES CCCCCCCCSCCCCCCCC
IUPAC Name 1-octylsulfanyloctane
Molecular Weight (g/mol) 258.51
PubChem CID 75901
Formula Weight 152.62
Percent Purity 95%
Chemical Name or Material Cinnamyl chloride, stabilized with sodium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.