missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl Bromide, TRC

Product Code. 30353766
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30353766 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30353766

Brand: Toronto Research Chemicals TRCC4420352.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinnamyl Bromide
CAS 4392-24-9
Molecular Formula C9H9Br
Quantity 2.5 g
Synonym 1-Bromo-3-phenyl-2-propene, (3-Bromo-1-propenyl)benzene, (3-Bromo-1-propen-1-yl)benzene, 1-Phenyl-3-bromo-1-propene, 3-Bromo-1-phenylprop-1-ene, 3-Bromo-1-phenylpropene, 3-Bromoprop-1-en-1-ylbenzene, 3-Phenyl-2-propenyl Bromide3-Phenylallyl Bromide, Cinnamyl Bromide, NSC 78446
InChI Formula InChI=1S/C9H9Br/c10-8-4-7-9-5-2-1-3-6-9/h1-7H,8H2/b7-4+
SMILES BrC\C=C\c1ccccc1
IUPAC Name [(E)-3-bromoprop-1-enyl]benzene
Molecular Weight (g/mol) 197.07
Formula Weight 195.9888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.