missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl Bromide, TRC

Product Code. 30355281
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30355281 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355281 Supplier Toronto Research Chemicals Supplier No. TRCC442035250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinnamyl Bromide
CAS 4392-24-9
Molecular Formula C9H9Br
Quantity 250 mg
Synonym 1-Bromo-3-phenyl-2-propene, (3-Bromo-1-propenyl)benzene, (3-Bromo-1-propen-1-yl)benzene, 1-Phenyl-3-bromo-1-propene, 3-Bromo-1-phenylprop-1-ene, 3-Bromo-1-phenylpropene, 3-Bromoprop-1-en-1-ylbenzene, 3-Phenyl-2-propenyl Bromide3-Phenylallyl Bromide, Cinnamyl Bromide, NSC 78446
InChI Formula InChI=1S/C9H9Br/c10-8-4-7-9-5-2-1-3-6-9/h1-7H,8H2/b7-4+
SMILES BrC\C=C\c1ccccc1
IUPAC Name [(E)-3-bromoprop-1-enyl]benzene
Molecular Weight (g/mol) 197.07
Formula Weight 195.9888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.