missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamonitrile, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Cinnamonitrile |
| CAS | 1885-38-7 |
| Molecular Formula | C9H7N |
| Quantity | 5 g |
| Synonym | (2E)-3-phenyl-2-propenenitrile, (2E)-3-Phenylprop-2-enenitrile, trans-3-Phenyl-2-propenenitrile, trans-3-Phenylpropenonitrile, trans-Cinnamonitrile, trans-beta-Phenylacrylonitrile |
| InChI Formula | InChI=1S/C9H7N/c10-8-4-7-9-5-2-1-3-6-9/h1-7H/b7-4+ |
| SMILES | N#C\C=C\c1ccccc1 |
| IUPAC Name | (E)-3-phenylprop-2-enenitrile |
| Molecular Weight (g/mol) | 129.16 |
| Formula Weight | 129.0578 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction