missing translation for 'onlineSavingsMsg'
Learn More

Cinnamonitrile, 97%, predominantly trans

Product Code. 10406720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10406720 25 g Glass bottle 25g
10098811 5 g Glass bottle 5g
10765941 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10406720 Supplier Thermo Scientific Acros Supplier No. 110380250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1885-38-7 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1885-38-7
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00001930
InChI Key ZWKNLRXFUTWSOY-QPJJXVBHSA-N
Synonym cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
PubChem CID 1550846
IUPAC Name (E)-3-phenylprop-2-enenitrile
SMILES C1=CC=C(C=C1)C=CC#N

Specifications

Melting Point 20°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 254°C to 255°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 10% max. (cis-Cinnamonitrile) (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCN
Refractive Index 1.6000 to 1.602
Quantity 25 g
Beilstein 09, 589
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble
Formula Weight 129.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cinnamonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Cinnamonitrile, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.