missing translation for 'onlineSavingsMsg'
Learn More

Cinnamaldehyde, TRC

Product Code. 18092230
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18092230 50 g 50g
18011740 10 g 10g
18001540 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092230 Supplier Toronto Research Chemicals Supplier No. TRCC44200550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-55-2
Molecular Formula C9 H8 O
Molecular Weight (g/mol) 132.16
Synonym Cinnamaldehyde (8CI), 3-Phenyl-2-propenal, 3-Phenyl-2-propen-1-al, 3-Phenyl-2-propenaldehyde, 3-Phenyl-2-propene-1-al, 3-Phenylacrolein, 3-Phenylacrylaldehyde, 3-Phenylpropenal, Abion CA, Benzylideneacetaldehyde, Cassia aldehyde, Cinnacure, Cinnamal, Cinnamic aldehyde, Cinnamite, Cinnamyl aldehyde, NSC 16935, NSC 40346, Phenylacrolein, XC 800, Zimtaldehyde, β-Phenylacrolein
IUPAC Name 3-phenylprop-2-enal
SMILES O=CC=Cc1ccccc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H
Formula Weight 132.0575
Percent Purity >95%
Chemical Name or Material Cinnamaldehyde
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.