missing translation for 'onlineSavingsMsg'
Learn More

Cinnabarinic Acid, TRC

Product Code. 30138211
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138211 25 mg 25mg
30137065 10 mg 10mg
30003543 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138211 Supplier Toronto Research Chemicals Supplier No. TRCC44200025MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-59-7
Molecular Formula C14 H8 N2 O6
Molecular Weight (g/mol) 300.22
Synonym 3H-Phenoxazine-1,9-dicarboxylic acid, 2-amino-3-oxo- (6CI, 7CI, 8CI, 9CI, ACI), 2-Amino-3-oxo-3H-phenoxazine-1,9-dicarboxylic acid (ACI), Cinnabaric acid, Cinnabarinic acid, Cinnavalininic acid
IUPAC Name 2-amino-3-oxophenoxazine-1,9-dicarboxylic acid
SMILES NC1=C(C(=O)O)C2=Nc3c(OC2=CC1=O)cccc3C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H8N2O6/c15-10-6(17)4-8-12(9(10)14(20)21)16-11-5(13(18)19)2-1-3-7(11)22-8/h1-4H,15H2,(H,18,19)(H,20,21)
Formula Weight 300.0382
Percent Purity >95%
Chemical Name or Material Cinnabarinic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.