missing translation for 'onlineSavingsMsg'
Learn More

Cinitapride Tartrate, TRC

Product Code. 30005057
Change view
Click to view available options
Quantity:
2.5 mg
50 mg
100 mg
Unit Size:
100mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005057 2.5 mg 2.5mg
30007144 100 mg 100mg
30010945 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005057 Supplier Toronto Research Chemicals Supplier No. TRCC4419952.5MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96623-56-2
Molecular Formula C21 H30 N4 O4 . C4 H6 O6
Molecular Weight (g/mol) 552.57
Synonym Dihydroxybutanedioate, 4-Amino-N-[1-(3-cyclohexen-1-ylmethyl)-4-piperidinyl]-2-ethoxy-5-nitrobenzamide [R-(R*,R*)]-2,3-Dihydroxybutanedioate, Blaston, Cidine, 4-Amino-N-[1-(3-cyclohexen-1-ylmethyl)-4-piperidinyl]-2-ethoxy-5-nitrobenzamide (2R,3R)-2,3-dihydroxybutanedioate (1:1)
IUPAC Name 4-amino-N-[1-(cyclohex-3-en-1-ylmethyl)-4-piperidyl]-2-ethoxy-5-nitro-benzamide;(2R,3R)-2,3-dihydroxybutanedioic acid
SMILES CCOc1cc(N)c(cc1C(=O)NC2CCN(CC3CCC=CC3)CC2)[N+](=O)[O-].O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C21H30N4O4.C4H6O6/c1-2-29-20-13-18(22)19(25(27)28)12-17(20)21(26)23-16-8-10-24(11-9-16)14-15-6-4-3-5-7-15;5-1(3(7)8)2(6)4(9)10/h3-4,12-13,15-16H,2,5-11,14,22H2,1H3,(H,23,26);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1
Formula Weight 552.2431
Percent Purity >95%
Chemical Name or Material Cinitapride Tartrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.