missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cingulin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21369-1-AP-20UL
This item is not returnable.
View return policy
Description
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predomitly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human maligcies, including several Acute Myeloid Leukemia studies.Specifications
| Cingulin | |
| Polyclonal | |
| Unconjugated | |
| CGN | |
| CGN, Cingulin, KIAA1319 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 310655, 57530, 70737 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59242, Q9P2M7 | |
| Cgn | |
| Cingulin Fusion Protein Ag15843 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction