missing translation for 'onlineSavingsMsg'
Learn More

Cinepazide Maleate, MedChemExpress

Product Code. 16454067
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16454067

Brand: Medchem Express HY66010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cinepazide Maleate (MD-67350) is a piperazine derivative and acts as a weak calcium channel blocker. Cinepazide Maleate is a potent vasodilator and can be used for the research of cerebrovascular diseases, including ischemic stroke, brain infarct et. al.

Cinepazide Maleate (MD-67350) is a piperazine derivative and acts as a weak calcium channel blocker. Cinepazide Maleate is a potent vasodilator and can be used for the research of cerebrovascular diseases, including ischemic stroke, brain infarct et. al.

Specifications

Chemical Name or Material Cinepazide Maleate
CAS 26328-04-1
Color White
Purity Grade Notes Research
Molecular Formula C26H35N3O9
Quantity 50 mg
Synonym MD-67350
Solubility Information DMSO : 100 mg/mL (187.42 mM; Need ultrasonic) ∣H2O : 50 mg/mL (93.71 mM; Need ultrasonic)
SMILES O=C(N1CCN(CC(N2CCCC2)=O)CC1)/C=C/C3=CC(OC)=C(OC)C(OC)=C3.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 533.57
Formula Weight 533.57
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.