missing translation for 'onlineSavingsMsg'
Learn More

Cineole, Approx. 99%, MP Biomedicals™

Product Code. 11442052 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442052 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442052 Supplier MP Biomedicals Supplier No. 0215068950
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Pharmaceuticals, Insecticide

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Color Colorless
Density 0.921 to 0.924g/mL
Assay Percent Range Approx. 99%
Quantity 50 mL
Formula Weight 154.3
Physical Form Liquid
Chemical Name or Material Cineole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.