missing translation for 'onlineSavingsMsg'
Learn More

1,8-Cineole, 99%

Product Code. p-3757310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
Product Code. Quantity Packaging unitSize
10695522 5 g Glass bottle 5g
10605532 100 g Glass bottle 100g
10588883 500 g Glass bottle 500g
3 options
This item is not returnable. View return policy

Product Code. 10695522

Brand: Thermo Scientific Acros 110340050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 470-82-6 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Melting Point 1.5°C
Color Colorless to Yellow
Density 0.9225g/mL
Boiling Point 176°C to 177°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.46
Quantity 5 g
Beilstein 17,II,32
Merck Index 15, 3938
Specific Gravity 0.9225
Specific Rotation -0.3° to +0.3° (20°C, 589nm) (neat)
Solubility Information (in ethanol (70% v/v)) 1:2
Formula Weight 154.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,8-Cineole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.