missing translation for 'onlineSavingsMsg'
Learn More

Cinchophen, LoGiCal

Product Code. 30461413
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461413 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461413 Supplier LoGiCal Supplier No. LGCFOR1978.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinchophen
CAS 132-60-5
Analyte or Component Names Cinchophen
Shipping Condition Room Temperature
Molecular Formula C16 H11 N O2
Certifications/Compliance ISO 9001
Synonym 4-Quinolinecarboxylic acid, 2-phenyl-, Cinchoninic acid, 2-phenyl- (8CI), 2-Phenyl-4-quinolinecarboxylic acid, 2-Phenylcinchonic acid, 2-Phenylcinchoninic acid, 4-Carboxy-2-phenylquinoline, Aciphenochinoline, Aciphenochinolinium, Agotan, Alutyl, Alutyo, Aminophan, Artam, Artexin, Atigoa, Atocin, Atofan, Atophan, Cinchophen, Cinchophene, Cinchophenic acid, Cinconal, Cincophen, Cincosal, Ikterosan, Mylofanol, NSC 2617, NSC 70178, Phenophan, Phenoquin, Polyphlogin, Quinofen, Quinophan, Quinophen, Rhematan, Rheumatan, Rheumin, Rheumine, Tervalon, Tophol, Tophosan, Traubofan, Vantyl, Viophan
InChI Formula InChI=1S/C16H11NO2/c18-16(19)13-10-15(11-6-2-1-3-7-11)17-14-9-5-4-8-12(13)14/h1-10H,(H,18,19)
Solution Type Neat
SMILES OC(=O)c1cc(nc2ccccc12)c3ccccc3
IUPAC Name 2-phenylquinoline-4-carboxylic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 249.26
Quantity 10 mg
Formula Weight 249.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.