missing translation for 'onlineSavingsMsg'
Learn More

Cinchonine, MedChemExpress

Product Code. 16409416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409416 Supplier Medchem Express Supplier No. HYY0152

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cinchonine is a natural compound present in Cinchona bark. Cinchonine activates endoplasmic reticulum stress-induced apoptosis in human liver cancer cells.

Cinchonine is a natural compound present in Cinchona bark. Cinchonine activates endoplasmic reticulum stress-induced apoptosis in human liver cancer cells.

Specifications

Chemical Name or Material Cinchonine
CAS 118-10-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H22N2O
Quantity 100 mg
Synonym (8R,9S)-Cinchonine LA40221
Solubility Information DMSO : 4.76 mg/mL (16.17 mM; Need ultrasonic)
SMILES O[C@@H](C1=CC=NC2=CC=CC=C12)[C@]3([H])[N@@]4C[C@H](C=C)[C@@H](CC4)C3
Molecular Weight (g/mol) 294.39
Formula Weight 294.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.