missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cinchonine, 98+%

Product Code. 10419930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10419930 100 g Glass bottle 100g
10696992 25 g Glass bottle 25g
11956621 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10419930 Supplier Thermo Scientific Chemicals Supplier No. 227811000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-10-5 | C19H22N2O | 294.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-10-5
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00064372
InChI Key KMPWYEUPVWOPIM-FRYPIZGFNA-N
Synonym +-quinolin-4-yl 5-vinylquinuclidin-2-yl methanol, 5r-5-vinylquinuclidin-2-yl 1s-4-quinolylmethan-1-ol
PubChem CID 21862290
IUPAC Name [(5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
SMILES [H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C

Specifications

Melting Point 255.0°C to 265.0°C
Color White to Beige
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Beilstein 23, V,12, 406
Fieser 06,501; 13,264
Merck Index 15,229
Specific Rotation Condition +251.00 (20.00°C c=2,0.2M HCl)
Specific Rotation + 251.00
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/60mL alcohol - 1g/25mL boiling alcohol, 1g/110mL chloroform - 1g/500mL ether
Formula Weight 294.4
Percent Purity 99%
Physical Form Micro-crystalline Powder
Chemical Name or Material Cinchonine
Show More Show Less
compliance-icons
Product Identifier
  • Cinchonine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.