missing translation for 'onlineSavingsMsg'
Learn More

Cinchonidine, MedChemExpress

Product Code. 16479016
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479016 Supplier Medchem Express Supplier No. HYN0173

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cinchonidine (α-Quinidine) is a cinchona alkaloid found in Cinchona officinalis and Gongronema latifolium. A building block used in asymmetric synthesis in organic chemistry. Weak inhibitor of serotonin transporter (SERT) with Kis of 330, 4.2, 36, 196, 15 μM for dSERT, hSERT, hSERT I172M, hSERT S438T, hSERT Y95F, respectively. Antimalarial activities.

Cinchonidine (α-Quinidine) is a cinchona alkaloid found in Cinchona officinalis and Gongronema latifolium. A building block used in asymmetric synthesis in organic chemistry. Weak inhibitor of serotonin transporter (SERT) with Kis of 330, 4.2, 36, 196, 15 μM for dSERT, hSERT, hSERT I172M, hSERT S438T, hSERT Y95F, respectively. Antimalarial activities.

Specifications

Chemical Name or Material Cinchonidine
Physical Form Powder
CAS 485-71-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H22N2O
For Use With (Application) COVID-19-immunoregulation
Synonym α-Quinidine
Solubility Information DMSO : 33.33 mg/mL (113.22 mM; Need ultrasonic)
SMILES C=C[C@H]1C[N@](CC[C@H]1C2)[C@]2([H])[C@H](O)C3=CC=NC4=CC=CC=C34
Molecular Weight (g/mol) 294.39
Formula Weight 294.39
Percent Purity 97.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.