missing translation for 'onlineSavingsMsg'
Learn More

(-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine

Product Code. 11473078
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473078 100 g 100g
11463078 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473078 Supplier Thermo Scientific Alfa Aesar Supplier No. A18796.22
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 485-71-2 | C19H22N2O | 294.40 g/mol

It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

Solubility
Insoluble in water. Soluble in alcohol and chloroform.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-71-2
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00006783
InChI Key KMPWYEUPVWOPIM-UHFFFAOYNA-N
Synonym cinchonidine
PubChem CID 45358337
SMILES OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 201°C to 206°C
Quantity 100 g
Beilstein 89690
Sensitivity Light sensitive
Merck Index 14,2286
Solubility Information Insoluble in water. Soluble in alcohol and chloroform.
Optical Rotation −109° (c=0.5 in Ethanol)
IUPAC Name [(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
Formula Weight 294.39
Percent Purity 99%
Assay (total base), may cont. up to 5% quinine
Chemical Name or Material (-)-Cinchonidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.