missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cinchonidine, 98.5-101%

Product Code. 10132330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10132330 25 g 25g
10755751 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10132330 Supplier Thermo Scientific Chemicals Supplier No. 110330250
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
This item is not returnable. View return policy

Cinchonidine, CAS # 485-71-2, is a naturally occurring alkaloid that may be used in asymmetric synthesis and exhibits anti-malarial activity | CAS: 485-71-2 | C19H22N2O | 294.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cinchonidine is an alkaloid naturally occurring in the bark of Cinchona strubs that has activity against Plasmodium spp
  • Cinchonidine is a diastereoisomer of cinchonine and can be used to promote chirality in asymmetric or enantioselective synthesis of organic compounds

Applications

  • Cinchonidine has been used as a starting material in the synthesis of anti-malarial cinchona alkaloid conjugates
  • Has been shown to potently induce γ-globin mRNA and fetal hemoglobin in erythroid progenitor cells and may be useful in preclinical studies of β-thalassemia
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-71-2
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00006783
InChI Key KMPWYEUPVWOPIM-UHFFFAOYNA-N
Synonym cinchonidine
PubChem CID 45358337
IUPAC Name [(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
SMILES OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 200.0°C to 203.0°C
Color White to Cream
Loss on Drying 1% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5-101%
Packaging Glass bottle
Quantity 25 g
Beilstein 23, 420
Merck Index 15, 2289
Specific Rotation − 115.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform
Formula Weight 294.39
Percent Purity 98.5-101%
Physical Form Crystalline Powder
Chemical Name or Material Cinchonidine, 98.5-101%
Show More Show Less
compliance-icons
Product Identifier
  • Cinchonidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: Cinchonidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.