missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinchocaine Hydrochloride, Mikromol™
Description
Cinchocaine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Cinchocaine hydrochloride |
| CAS | 61-12-1 |
| Analyte or Component Names | Cinchocaine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H29 N3 O2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cinchocaine Hydrochloride, 2-Butoxy-N-[2-(diethylamino)ethyl]-4-quinolinecarboxamide monohydrochloride, 2-Butoxy-N-[2-(diethylamino)ethyl]-cinchoninamide monohydrochloride, 2-Butoxy-N-(2-diethylaminoethyl)cinchoninamide hydrochloride, 2-Butoxy-N-(2-diethylaminoethyl)cinchoninic acid amide hydrochloride, 2-n-Butoxy-N-(2-diethylaminoethyl)cinchoninamide hydrochloride, Benzolin, Benzolin (local anesthetic), Cincaine, Cincaine chloride, Cinchocainium chloride, Dibucaine hydrochloride, Nupercaine hydrochloride, Percain, Percaine, Percamin S, Sovcain, Sovcaine, 4-Quinolinecarboxamide, 2-butoxy-N-[2-(diethylamino)ethyl]-, monohydrochloride (9CI), Cinchoninamide, 2-butoxy-N-[2-(diethylamino)ethyl]-, monohydrochloride (8CI), Cinchocaine hydrochloride |
| InChI Formula | InChI=1S/C20H29N3O2.ClH/c1-4-7-14-25-19-15-17(16-10-8-9-11-18(16)22-19)20(24)21-12-13-23(5-2)6-3;/h8-11,15H,4-7,12-14H2,1-3H3,(H,21,24);1H |
| SMILES | Cl.CCCCOc1cc(C(=O)NCCN(CC)CC)c2ccccc2n1 |
| IUPAC Name | 2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?