missing translation for 'onlineSavingsMsg'
Learn More

Cinchocaine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30460910
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460910 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460910 Supplier LoGiCal Supplier No. LGCAMP0604.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinchocaine hydrochloride
CAS 61-12-1
Analyte or Component Names Cinchocaine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H29 N3 O2 . Cl H
Certifications/Compliance ISO 17034
Synonym Cinchocaine Hydrochloride, 2-Butoxy-N-[2-(diethylamino)ethyl]-4-quinolinecarboxamide monohydrochloride, 2-Butoxy-N-[2-(diethylamino)ethyl]-cinchoninamide monohydrochloride, 2-Butoxy-N-(2-diethylaminoethyl)cinchoninamide hydrochloride, 2-Butoxy-N-(2-diethylaminoethyl)cinchoninic acid amide hydrochloride, 2-n-Butoxy-N-(2-diethylaminoethyl)cinchoninamide hydrochloride, Benzolin, Benzolin (local anesthetic), Cincaine, Cincaine chloride, Cinchocainium chloride, Dibucaine hydrochloride, Nupercaine hydrochloride, Percain, Percaine, Percamin S, Sovcain, Sovcaine, 4-Quinolinecarboxamide, 2-butoxy-N-[2-(diethylamino)ethyl]-, monohydrochloride (9CI), Cinchoninamide, 2-butoxy-N-[2-(diethylamino)ethyl]-, monohydrochloride (8CI), Cinchocaine hydrochloride
InChI Formula InChI=1S/C20H29N3O2.ClH/c1-4-7-14-25-19-15-17(16-10-8-9-11-18(16)22-19)20(24)21-12-13-23(5-2)6-3;/h8-11,15H,4-7,12-14H2,1-3H3,(H,21,24);1H
Solution Type Single Solution
SMILES Cl.CCCCOc1cc(C(=O)NCCN(CC)CC)c2ccccc2n1
IUPAC Name 2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 379.92
Quantity 1 mL
Formula Weight 379.2027 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.