missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CINAP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92107-0.1ml
This item is not returnable.
View return policy
Description
CINAP Polyclonal antibody specifically detects CINAP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CINAP | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| CASK-interacting nucleosome assembly protein, CDA1CTCL-associated antigen se20-4, Cell division autoantigen 1, CINAPSE20-4, CTCL, CTCL tumor antigen se20-4, Cutaneous T-cell lymphoma-associated antigen se20-4, cutaneous T-cell lymphoma-associated tumor antigen se20-4, DENTTTSPY-like protein 2, differentially expressed nucleolar TGF-beta1 target, Differentially-expressed nucleolar TGF-beta1 target protein, HRIHFB2216, NP79, Nuclear protein of 79 kDa, SE204, testis-specific protein Y encoded-like 2, testis-specific Y-encoded-like protein 2, TSPX, TSPY-like 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 240-460 of human TSPYL2 (NP_071400.1). LKRKFIQMRRPFLERRDLIIQHIPGFWVKAFLNHPRISILINRRDEDIFRYLTNLQVQDLRHISMGYKMKLYFQTNPYFTNMVIVKEFQRNRSGRLVSHSTPIRWHRGQEPQARRHGNQDASHSFFSWFSNHSLPEADRIAEIIKNDLWVNPLRYYLRERGSRIKRKKQEMKKRKTRGRCEVVIMEDAPDYYAVEDIFSEISDIDETIHDIKISDFMETTD | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Neuroscience | |
| 64061 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction