missing translation for 'onlineSavingsMsg'
Learn More

Cinacalcet hydrochloride, MedChemExpress

Product Code. p-200096412
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16497926

Brand: Medchem Express HY70037A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of Ca receptor (CaR), used for cardiovascular disease treatment.

Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of Ca receptor (CaR), used for cardiovascular disease treatment.

Specifications

Chemical Name or Material Cinacalcet hydrochloride
Physical Form Solid
CAS 364782-34-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H23ClF3N
For Use With (Application) COVID-19-immunoregulation
Synonym AMG-073 hydrochloride
Solubility Information DMSO : ≥ 50 mg/mL (126.95 mM) ∣H2O : 1 mg/mL (2.54 mM; ultrasonic and warming and heat to 60°C)
SMILES FC(F)(C1=CC(CCCN[C@@H](C2=C3C=CC=CC3=CC=C2)C)=CC=C1)F.Cl
Molecular Weight (g/mol) 393.87
Formula Weight 393.87
Percent Purity 99.73%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.