missing translation for 'onlineSavingsMsg'
Learn More

Cinacalcet hydrochloride, MedChemExpress

Product Code. 16497926
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497926 100 mg 100mg
16484077 50 mg 50mg
16448178 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497926 Supplier Medchem Express Supplier No. HY70037A
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of Ca receptor (CaR), used for cardiovascular disease treatment.

Cinacalcet hydrochloride (AMG-073 hydrochloride) is an orally active, allosteric agonist of Ca receptor (CaR), used for cardiovascular disease treatment.

Specifications

Chemical Name or Material Cinacalcet hydrochloride
Physical Form Solid
CAS 364782-34-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H23ClF3N
For Use With (Application) COVID-19-immunoregulation
Synonym AMG-073 hydrochloride
Solubility Information DMSO : ≥ 50 mg/mL (126.95 mM) ∣H2O : 1 mg/mL (2.54 mM; ultrasonic and warming and heat to 60°C)
SMILES FC(F)(C1=CC(CCCN[C@@H](C2=C3C=CC=CC3=CC=C2)C)=CC=C1)F.Cl
Molecular Weight (g/mol) 393.87
Formula Weight 393.87
Percent Purity 99.73%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.