missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinacalcet Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Cinacalcet Hydrochloride |
| CAS | 364782-34-3 |
| Analyte or Component Names | Cinacalcet Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H22 F3 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | AMG 073 HCl, Cinacalcet hydrochloride, KRN 1493, Mimpara, N-[(1R)-1-(1-Naphthyl)ethyl]-3-[3-(trifluoromethyl)phenyl]propan-1-amine hydrochloride, Regpara, Sensipar, 1-Naphthalenemethanamine, alpha-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride (1:1), (alphaR)-, 1-Naphthalenemethanamine, alpha-methyl-N-[3-[3-(trifluoromethyl)phenyl]propyl]-, hydrochloride, (alphaR)- (9CI) |
| InChI Formula | InChI=1S/C22H22F3N.ClH/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25;/h2-5,7,9-13,15-16,26H,6,8,14H2,1H3;1H/t16-;/m1./s1 |
| Solution Type | Neat |
| SMILES | Cl.C[C@@H](NCCCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?