missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cinacalcet hydrochloride

Product Code. 18736258
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18736258 50 mg 50mg
18734739 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18736258 Supplier R&D Systems™ Supplier No. 6170/50
  • ONLINE EXCLUSIVE PRICE
    £412.31
    £369.60 / 50mg
    Save £42.71
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Low molecular weight, organic compound that may help regulate biological process.

Cinacalcet hydrochloride is a calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

CAS 364782-34-3
Quantity 50 mg
Product Line Tocris
Target Calcium-Sensing Receptor
Molecular Formula C22H22F3N.HCl
Formula Weight 393.87g/mol
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.