missing translation for 'onlineSavingsMsg'
Learn More

Cimifugin, MedChemExpress

Product Code. p-200099131
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16459048

Brand: Medchem Express HYN0634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cimifugin (Cimitin) is a bioactive component of Saposhnikovia divaricata, a Chinese herb. Cimifugin suppresses allergic inflammation by reducing epithelial derived initiative key factors via regulating tight junctions.

Cimifugin (Cimitin) is a bioactive component of Saposhnikovia divaricata, a Chinese herb. Cimifugin suppresses allergic inflammation by reducing epithelial derived initiative key factors via regulating tight junctions.

Specifications

Chemical Name or Material Cimifugin
CAS 37921-38-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H18O6
Quantity 10 mg
Synonym Cimitin
Solubility Information DMSO : 100 mg/mL (326.47 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (108.81 mM; Need ultrasonic)
SMILES O=C1C=C(CO)OC2=CC(O[C@H](C(C)(O)C)C3)=C3C(OC)=C12
Molecular Weight (g/mol) 306.31
Formula Weight 306.31
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.