missing translation for 'onlineSavingsMsg'
Learn More

Cimetropium Bromide, MedChemExpress

Product Code. 16483149
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16483149

Brand: Medchem Express HYU00106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cimetropium Bromide (DA-3177) is a mAChR antagonist for long-term treatment of irritable bowel syndrome.

Cimetropium Bromide (DA-3177) is a mAChR antagonist for long-term treatment of irritable bowel syndrome.

Specifications

Chemical Name or Material Cimetropium Bromide
CAS 51598-60-8
Color White
Purity Grade Notes Research
Molecular Formula C21H28BrNO4
Quantity 1 mg
Synonym DA-3177
Solubility Information H2O : 50 mg/mL (114.06 mM; Need ultrasonic)
SMILES C[N+]1([C@H]2[C@@H](O3)[C@@H]3[C@@H]1C[C@H](OC([C@@H](C4=CC=CC=C4)CO)=O)C2)CC5CC5.[Br-]
Molecular Weight (g/mol) 438.36
Formula Weight 438.36
Percent Purity 95.34%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.