missing translation for 'onlineSavingsMsg'
Learn More

Cimetidine, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001143
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30001143

Brand: Tocris Bioscience 0902/100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

H2 antagonist, I1 agonist

Widely used H2 histamine antagonist which has more recently been described as an inverse agonist. Also a potent I1 imidazoline binding site ligand.
TRUSTED_SUSTAINABILITY

Specifications

CAS 51481-61-9
Molecular Formula C10H16N6S
Quantity 100 mg
Synonym cimetidine, tagamet, tametin, eureceptor, cimetag, acinil, tratul, cimetidina, dyspamet, ulcedine
InChI Key AQIXAKUUQRKLND-UHFFFAOYSA-N
SMILES CC1=C(N=CN1)CSCCNC(=NC)NC#N
IUPAC Name 1-cyano-2-methyl-3-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]guanidine
Molecular Weight (g/mol) 252.34
PubChem CID 2756
ChEBI CHEBI:3699
Formula Weight 252.34
Percent Purity >99%
Physical Form Solid
Chemical Name or Material Cimetidine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.