missing translation for 'onlineSavingsMsg'
Learn More

Cimetidine, MedChemExpress

Product Code. 16431409
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 16431409

Brand: Medchem Express HY14289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cimetidine (SKF-92334) is an orally active and inverse histamine H2 receptor antagonist with a Ki of 0.6 μM. Cimetidine is a gastric acid reducer, and can be used for duodenal and gastric ulcers research. Cimetidine has anti-cancer and anti-inflammatory activity.

Cimetidine (SKF-92334) is an orally active and inverse histamine H2 receptor antagonist with a Ki of 0.6 μM. Cimetidine is a gastric acid reducer, and can be used for duodenal and gastric ulcers research. Cimetidine has anti-cancer and anti-inflammatory activity.

Specifications

Chemical Name or Material Cimetidine
Physical Form Solid
CAS 51481-61-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C10H16N6S
For Use With (Application) Cancer-programmed cell death
Synonym SKF-92334
Solubility Information DMSO : 60 mg/mL (237.77 mM; Need ultrasonic) ∣H2O : 2 mg/mL (7.93 mM; Need ultrasonic)
SMILES CC1=C(CSCC/N=C(NC)/NC#N)NC=N1
Molecular Weight (g/mol) 252.34
Formula Weight 252.34
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.