missing translation for 'onlineSavingsMsg'
Learn More

CIM 0216, TRC

Product Code. 17951765
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951765 10 mg 10mg
17910745 5 mg 5mg
17921345 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17951765 Supplier Toronto Research Chemicals Supplier No. TRCC44155010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1031496-06-6
Molecular Formula C21 H21 N3 O2
Molecular Weight (g/mol) 347.41
Synonym 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide, CIM 0216
IUPAC Name 2-(3,4-dihydro-2H-quinolin-1-yl)-N-(5-methylisoxazol-3-yl)-2-phenyl-acetamide
SMILES Cc1onc(NC(=O)C(N2CCCc3ccccc23)c4ccccc4)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H21N3O2/c1-15-14-19(23-26-15)22-21(25)20(17-9-3-2-4-10-17)24-13-7-11-16-8-5-6-12-18(16)24/h2-6,8-10,12,14,20H,7,11,13H2,1H3,(H,22,23,25)
Formula Weight 347.1634
Chemical Name or Material 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-alpha-phenyl-1(2H)-quinolineacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.