missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CILP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559753
This item is not returnable.
View return policy
Description
Immunogen sequence: LRFARILLGQ EPIGFTAYQG DFTIEVPPST QRLVVTFVDP SGEFMDAVRV LPFDPRGAGV YHEVKAMRKK APVILHTSQS NTIPLGELED EAPLGELVLP SGAFRRADGK PYSGP Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.
Specifications
| CILP2 | |
| Polyclonal | |
| Unconjugated | |
| CILP2 | |
| 1110031K21Rik; AA407300; cartilage intermediate layer protein 2; Cartilage intermediate layer protein 2 C1; Cartilage intermediate layer protein 2 C2; cartilage intermediate layer protein-like protein 2; Cilp2; CILP-2; CLIP-2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 148113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.50 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUL8 | |
| CILP2 | |
| Recombinant protein corresponding to Human CILP2. Recombinant protein control fragment (Product #RP-99119). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction