missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CILP-1 Recombinant Protein Antigen
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantity:
0.1 mL
Dimensione della confezione:
0.1mL
Descrizione
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CILP. The CILP-1 Recombinant Protein Antigen is derived from E. coli. The CILP-1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-81667. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Specifica
Specifica
| Gene ID (Entrez) | 8483 |
| Species | Human |
| Purification Method | Chromatography |
| Purity | >80% |
| Concentration | 0.5mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Symbol | CILP |
| Label Type | Unlabeled |
| Vedi altri risultati |
For Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto