missing translation for 'onlineSavingsMsg'
Learn More

Cilostamide, Tocris Bioscience™

Catalog Number p-4783350
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
68550-75-4
C20H26N2O3
342.439
UIAYVIIHMORPSJ-UHFFFAOYSA-N
cilostamide, ciloalamide, cilostamide inn, cilostamidum inn-latin, cilostamida inn-spanish, n-cyclohexyl-n-methyl-4-1,2-dihydro-2-oxo-6-quinolyloxy butyramide, chembl34431, n-cyclohexyl-n-methyl-4-2-oxo-1h-quinolin-6-yl oxy butanamide, butanamide, n-cyclohexyl-4-1,2-dihydro-2-oxo-6-quinolinyl oxy-n-methyl, dsstox_cid_25140
2753
N-cyclohexyl-N-methyl-4-[(2-oxo-1H-quinolin-6-yl)oxy]butanamide
CN(C1CCCCC1)C(=O)CCCOC2=CC3=C(C=C2)NC(=O)C=C3
This item is not returnable. View return policy
68550-75-4
C20H26N2O3
342.439
UIAYVIIHMORPSJ-UHFFFAOYSA-N
cilostamide, ciloalamide, cilostamide inn, cilostamidum inn-latin, cilostamida inn-spanish, n-cyclohexyl-n-methyl-4-1,2-dihydro-2-oxo-6-quinolyloxy butyramide, chembl34431, n-cyclohexyl-n-methyl-4-2-oxo-1h-quinolin-6-yl oxy butanamide, butanamide, n-cyclohexyl-4-1,2-dihydro-2-oxo-6-quinolinyl oxy-n-methyl, dsstox_cid_25140
2753
N-cyclohexyl-N-methyl-4-[(2-oxo-1H-quinolin-6-yl)oxy]butanamide
CN(C1CCCCC1)C(=O)CCCOC2=CC3=C(C=C2)NC(=O)C=C3

PDE3 inhibitor

150Selective inhibitor of type III phosphodiesterase (PDE3). Displays moderate selectivity for PDE3A isozyme vs. PDE3B (IC50 values are 0.027 and 0.050 μM for PDE3A and PDE3B respectively). Inhibits ADP-induced platelet aggregation (IC50 = 16.8 μM); antithrombotic.
Quantity 50 mg
Formula Weight 342.44
Percent Purity >98%
Chemical Name or Material Cilostamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'