missing translation for 'onlineSavingsMsg'
Learn More

Cilostamide

Product Code. 15406839
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406839 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15406839 Supplier Thermo Scientific Alfa Aesar Supplier No. J65319.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68550-75-4 | C20H26N2O3 | 342.439 g/mol

A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 68550-75-4
Color White
Molecular Formula C20H26N2O3
MDL Number MFCD00673958
Quantity 10 mg
Synonym cilostamide, ciloalamide, cilostamide inn, cilostamidum inn-latin, cilostamida inn-spanish, n-cyclohexyl-n-methyl-4-1,2-dihydro-2-oxo-6-quinolyloxy butyramide, chembl34431, n-cyclohexyl-n-methyl-4-2-oxo-1h-quinolin-6-yl oxy butanamide, butanamide, n-cyclohexyl-4-1,2-dihydro-2-oxo-6-quinolinyl oxy-n-methyl, dsstox_cid_25140
InChI Key UIAYVIIHMORPSJ-UHFFFAOYSA-N
SMILES CN(C1CCCCC1)C(=O)CCCOC2=CC3=C(C=C2)NC(=O)C=C3
IUPAC Name N-cyclohexyl-N-methyl-4-[(2-oxo-1H-quinolin-6-yl)oxy]butanamide
Molecular Weight (g/mol) 342.439
PubChem CID 2753
Formula Weight 342.43
Physical Form Powder
Chemical Name or Material Cilostamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.