missing translation for 'onlineSavingsMsg'
Learn More

Cilostamide, MedChemExpress

Product Code. 16494576
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494576 100 mg 100mg
16489866 50 mg 50mg
16466057 5 mg 5mg
16442726 25 mg 25mg
16423338 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494576 Supplier Medchem Express Supplier No. HY101312
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cilostamide is a selective and potent PDE3 inhibitor, with IC50s of 27 nM and 50 nM for PDE3A and PDE3B, respectively, and has antithrombotic and anti-intimal hyperplastic activity.

Cilostamide is a selective and potent PDE3 inhibitor, with IC50s of 27 nM and 50 nM for PDE3A and PDE3B, respectively, and has antithrombotic and anti-intimal hyperplastic activity.

Specifications

Chemical Name or Material Cilostamide
CAS 68550-75-4
Color White
Purity Grade Notes Research
Molecular Formula C20H26N2O3
Quantity 100 mg
Synonym OPC3689
Solubility Information DMSO : 31 mg/mL (90.53 mM; Need ultrasonic and warming)
SMILES O=C(N(C1CCCCC1)C)CCCOC2=CC3=C(NC(C=C3)=O)C=C2
Molecular Weight (g/mol) 342.43
Formula Weight 342.43
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.