missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cilostamide

Product Code. 30001406
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001406 10 mg 10mg
18094932 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001406 Supplier R&D Systems™ Supplier No. 0915/10
  • ONLINE EXCLUSIVE PRICE
    £178.91
    £159.60 / 10mg
    Save £19.31
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PDE3 inhibitor

150Selective inhibitor of type III phosphodiesterase (PDE3). Displays moderate selectivity for PDE3A isozyme vs. PDE3B (IC50 values are 0.027 and 0.050 μM for PDE3A and PDE3B respectively). Inhibits ADP-induced platelet aggregation (IC50 = 16.8 μM); antithrombotic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68550-75-4
Molecular Formula C20H26N2O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 342.44
Purity >98%
Molecular Weight (g/mol) 342.439
Chemical Name or Material Cilostamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.