missing translation for 'onlineSavingsMsg'
Learn More

Cilofexor, MedChemExpress

Product Code. 16404278
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16404278

Brand: Medchem Express HY109083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cilofexor (GS-9674) is a potent, selective and orally active nonsteroidal FXR agonist with an EC50 of 43 nM. Cilofexor has anti-inflammatory and antifibrotic effects. Cilofexor has the potential for primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH) research.

Cilofexor (GS-9674) is a potent, selective and orally active nonsteroidal FXR agonist with an EC50 of 43 nM. Cilofexor has anti-inflammatory and antifibrotic effects. Cilofexor has the potential for primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH) research.

Specifications

Chemical Name or Material Cilofexor
CAS 1418274-28-8
Color White
Purity Grade Notes Research
Molecular Formula C28H22Cl3N3O5
Quantity 10 mg
Synonym GS-9674
Solubility Information DMSO : 50 mg/mL (85.20 mM; Need ultrasonic)
SMILES ClC1=C(C2(CN(C3=CC(C(O)=O)=CC=N3)C2)O)C=CC(OCC4=C(C5CC5)ON=C4C(C(Cl)=CC=C6)=C6Cl)=C1
Molecular Weight (g/mol) 586.85
Formula Weight 586.85
Percent Purity 98.25%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.