missing translation for 'onlineSavingsMsg'
Learn More

Cilnidipine, Tocris Bioscience™

Product Code. 18762634
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18762634 10 mg 10mg
18732594 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18762634 Supplier Tocris Bioscience Supplier No. 2629/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ca2+ channel blocker (dual L/N-type)

Dual L- and N-type calcium channel blocker that displays antihypertensive, sympatholytic and neuroprotective activity. Lowers mean blood pressure and reduces the size of cerebral infarction in the rat model of focal brain ischemia.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132203-70-4
Molecular Formula C27H28N2O7
Molecular Weight (g/mol) 492.528
InChI Key KJEBULYHNRNJTE-DHZHZOJOSA-N
Synonym cilnidipine, cinalong, atelec, siscard, cilnidipine inn, cinaldipine, 3-cinnamyl 5-2-methoxyethyl 2,6-dimethyl-4-3-nitrophenyl-1,4-dihydropyridine-3,5-dicarboxylate, atelec tn, +--e-cinnamyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate, 1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 2-methoxyethyl 2e-3-phenyl-2-propenyl ester
PubChem CID 5282138
ChEBI CHEBI:31399
IUPAC Name 3-O-(2-methoxyethyl) 5-O-[(E)-3-phenylprop-2-enyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCC=CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OCCOC

Specifications

Quantity 10 mg
Formula Weight 492.52
Percent Purity >99%
Chemical Name or Material Cilnidipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.